Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Rabbit IgG Fc, Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 568

Catalog No. A78955
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A78955 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A78955 Supplier Invitrogen™ Supplier No. A78955
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Secondary Antibody

This goat anti-rabbit IgG Fc secondary antibody has been affinity-purified and shows minimum cross-reactivity to human, mouse and rat serum proteins. These antibodies have been solid phase adsorbed to ensure class specificity and cross adsorbed using human, mouse and rat immunosorbents to remove cross reactive antibodies. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondary antibodies flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g. flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 568 dye is a bright, orange/red-fluorescent dye with excitation ideally suited to the 568 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 568 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abu...

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG Fc, Cross-Adsorbed
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 568
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Goat
Immunogen Gamma Immunoglobin Fc region.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.