Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Rabbit, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP187FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP187FMI 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP187FMI Supplier MilliporeSigma Supplier No. AP187F
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Goat anti-Rabbit IgG Antibody, FITC conjugate, Species Adsorbed detects level of Rabbit IgG and has been published and validated for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rabbit IgG Species adsorbed
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation Purified from immunoaffinity chromatography. Lyophilized from 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 15mg/mL Bovine Serum Albumin (Protease-Free, IgG-Free) and 0.05 Sodium Azide.
Host Species Goat
Purification Method Immunoaffinity chromatography
Quantity 1.5 mg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Protect from light
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.