Learn More
STEMCELL Technologies Goat Anti-Rabbit IgG (H+L) Antibody, Polyclonal, AMCA, Size: 1.5 mg
SDP
Supplier:  STEMCELL Technologies 60129AV
Encompass
This goat polyclonal antibody reacts with the heavy (H) chains on rabbit IgG and the light (L) chains common in most rabbit immunoglobulins. Use the polyclonal goat anti-rabbit IgG (H+L) antibody with applications, such as immunohistochemistry (IHC) and immunofluorescence (IF) assays. It has been verified for use with immunocytochemistry (ICC).
Catalog No. 50-197-5704