Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Anti-Rabbit IgG (H&L) Antibody (biotin), ProSci
SDP

Catalog No. 16817892
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-817-892 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-817-892 Supplier ProSci LLC Supplier No. XR99402000
Add to Cart
Add to Cart

Secondary Antibody

Goat Anti-Rabbit IgG (H&L) Antibody specifically detects Goat Anti-Rabbit IgG (H&L) in Rabbit Samples. It is validated for ELISA.

Specifications

Antigen Goat Anti-Rabbit IgG (H&L)
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Biotin Conjugated Affinity Purified anti-Rabbit IgG is suitable for immunoblotting, ELISA, immunohistochemistry, immunomicroscopy as well as other antibody based assay using streptavidin or avidin conjugates requiring lot-to-lot consistency. This product has been assayed against 1.0 ug of Rabbit IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiozoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature. A working dilution of 1:50,000 to 1:500,000 of the reconstitution concentration is suggested for this product. (Optimal dilution should be determined by user).
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, 0.01% (w/v) Sodium Azide; pH 7.2; 10 mg/mL Bovine Serum Albumin
Host Species Goat
Immunogen Rabbit IgG - whole molecule
Purification Method Biotin Conjugated Affinity Purified anti-Rabbit IgG was prepared from monospecific antiserum by immunoaffinity chromatography using Rabbit IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Goat Serum, Rabbit IgG and Rabbit Serum. No reaction was observed against Human Serum Proteins.
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Minimum Cross reactivity to Human Serum Proteins
Target Species Rabbit
Content And Storage Biotin Conjugated Affinity Purified anti-Rabbit IgG can be stored vial at 4 °C prior to restoration. For extended storage mix product with 50% glycerol and then aliquot contents and freeze at -20 °C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.