Learn More
Goat Anti-Rabbit IgG (H&L) Antibody (HRP), ProSci

Description
Specifications
Specifications
| Antigen | Goat Anti-Rabbit IgG (H&L) |
| Applications | ELISA, Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Peroxidase Conjugated Affinity Purified anti-Rabbit IgG is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency. Recommended dilutions are as follows, ELISA 1:20,000 - 1:100,000, Western Blot 1:2,000 - 1:10,000, Immunohistochemistry 1:1,000 - 1:5,000. (Optimal dilution should be determined by user). |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2; 10 mg/mL Bovine Serum Albumin (BSA); 0.01% (w/v) Gentamicin Sulfate. Do NOT add Sodium Azide! |
| Host Species | Goat |
| Immunogen | Rabbit IgG - whole molecule |
| Purification Method | Peroxidase Conjugated Affinity Purified anti-Rabbit IgG was prepared from monospecific antiserum by immunoaffinity chromatography using Rabbit IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Goat Serum, Rabbit IgG and Rabbit Serum. No reaction was observed against Human Serum Proteins. |
| Quantity | 2 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.