Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 488

Catalog No. PI35553 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI35553 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI35553 Supplier Invitrogen™ Supplier No. 35553
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Secondary Antibody

Product # 35553 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35553 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by solid-phase adsorbed to ensure minimal cross-reactivity with human, horse, mouse, swine, goat, and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 488 Amax= 493 nm; Emax= 518 nm. Mole Dye/Mole Protein Ratio is lot-dependent.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG (H+L) Cross-Adsorbed
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate DyLight 488
Formulation PBS with 0.02% sodium azide; pH 7.2
Host Species Goat
Purification Method Antigen affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.