Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, DyLighT&Trade; 488
Description
Product # 35552 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35552 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 488 Amax= 493 nm; Emax= 518 nm. Mole Dye/Mole Protein Ratio is lot-dependent.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | DyLight 488 |
| Formulation | PBS with 1% BSA and 0.02% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Purified Rabbit IgG, whole molecule. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.