Learn More
Cell Signaling Technology Secondary Antibody, Goat Anti-Rabbit IgG, Alexa Fluor™ 532 Conjugate, 250 μL
SDP
Catalog No. 503132410
Encompass
Goat Anti-Rabbit IgG (H™L) Antibody (Alexa Fluor™ 532 Conjugate) is a fluorescently labeled secondary antibody optimized for immunofluorescent applications. This antibody is prepared from goat and specifically detects both the heavy and light chains of rabbit IgG. It is cross-adsorbed against human, mouse, and bovine serum to ensure minimal cross-reactivity in multi-color staining experiments. The antibody is formulated at a concentration of 2 mg/mL and is suitable for high-performance flow cytometry analysis.
  • Excitation and emission wavelengths of 531 nm and 554 nm, respectively
  • Optimized for flow cytometry (fixed/permeabilized and live) applications
  • Cross-adsorbed against human IgG, human serum, mouse IgG, mouse serum, and bovine serum
  • Supplied in a stable formulation at 2 mg/mL
  • Validated for research use only
Catalog No. 50-313-2410 Supplier Cell Signaling Technology Supplier No. 74346S
Goat Anti-Rabbit IgG (H™L) Antibody (Alexa Fluor™ 532 Conjugate) is a fluorescently labeled secondary antibody optimized for immunofluorescent applications. This antibody is prepared from goat and specifically detects both the heavy and light chains of rabbit IgG. It is cross-adsorbed against human, mouse, and bovine serum to ensure minimal cross-reactivity in multi-color staining experiments. The antibody is formulated at a concentration of 2 mg/mL and is suitable for high-performance flow cytometry analysis.
  • Excitation and emission wavelengths of 531 nm and 554 nm, respectively
  • Optimized for flow cytometry (fixed/permeabilized and live) applications
  • Cross-adsorbed against human IgG, human serum, mouse IgG, mouse serum, and bovine serum
  • Supplied in a stable formulation at 2 mg/mL
  • Validated for research use only