Learn More
STEMCELL Technologies Goat Anti-Rabbit IgG (H+L) Antibody, Polyclonal, iFluor 568, Size: 200 ug
SDP
Supplier:  STEMCELL Technologies 1001083
Encompass
This goat polyclonal antibody reacts with the heavy (H) chains on rabbit IgG and the light (L) chains common in most rabbit immunoglobulins. Use the polyclonal goat anti-rabbit IgG (HL) antibody with applications such as immunohistochemistry (IHC) and immunofluorescence (IF) assays. It has been verified for use with immunocytochemistry (ICC).
Catalog No. 50-239-0243