Learn More
Bio Rad Laboratories GOAT ANTI-RABBIT IGG HRP 2ML
Supplier:  Bio Rad Laboratories 1706515
Encompass
Goat anti-rabbit IgG (H + L)-HRP is a goat secondary antibody conjugated with horseradish peroxidase (HRP) that is used to detect rabbit primary antibodies. Supplied as 2 ml conjugate. (170-6515)
Catalog No. NC9269486