Learn More
IgG Goat anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Goat anti-Rabbit IgG Species adsorbed |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8mg/mL |
| Conjugate | HRP |
| Formulation | Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA. RECONSTITUTION: Reconstitute with 1.5mLsterile distilled water. |
| Host Species | Goat |
| Quantity | 1.5 mL |
| Primary or Secondary | Secondary |
| Target Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.