Learn More
Bioworld GOAT ANTI-RABBIT IGGH+L)
Supplier:  Bioworld 201810812
Encompass
GOAT ANTI-RABBIT IGG (H+L) -RED SEPAROPORE (AGAROSE): Affinity isolated antibody goat anti-rabbit IgG coupled to cyanogen bromide-activated Red Separopore offers the advantage of increased sensitiveity for rabbit IgG without cross-reacti...
Catalog No. 50-197-877