Learn More
Jackson Immuno Research Labs GOAT ANTI-RABBIT RHODAMINE CON
SDP
Supplier:  Jackson Immuno Research Labs 111025144
Encompass_Preferred
Rhodamine (TRITC)-AffiniPure Goat Anti-Rabbit IgG (H+L) (min X Hu,Ms,Rat Sr Prot)
Catalog No. NC9748932
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov