Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat Ig, Polyclonal, FITC, Secondary Antibody, BD

Catalog No. BDB554016 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554016 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554016 Supplier BD Biosciences Supplier No. 554016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Host Species: Goat
Clone: Polyclonal
Isotype: Ig
Species Reactivity: Rat

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rat Ig
Classification Polyclonal
Clone POLYCLONAL
Concentration 0.5mg/mL
Conjugate FITC
Host Species Goat
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rat
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.