Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Rat, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP183CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP183CMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP183CMI Supplier MilliporeSigma Supplier No. AP183C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Goat anti-Rat IgG Antibody, Cy3 conjugate, Species Adsorbed is an antibody against Rat IgG for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rat IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Concentration 1.4mg/mL
Conjugate Cyanine3
Host Species Goat
Immunogen Highly purified Rat IgG isolated from pooled normal Mouse Serum.
Quantity 500 μg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C. Do not freeze.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.