Learn More
Goat anti-Rat IgG F(ab')2 fragment, R-PE, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Goat anti-Rat IgG F(ab')2 fragment |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | R-PE |
| Description | Goat anti-rat IgG recognizes rat IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Phycoerythrin. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilize from 10mM KH2PO4, 250mM NaCl, pH 7.2 (0.05% sodium azide) |
| Host Species | Goat |
| Immunogen | Rat IgG F(ab')2 fragment |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.