Learn More
IgG Goat anti-Rat, Phycoerythrin, Polyclonal, Secondary Antibody, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Goat anti-Rat IgG Fc Species adsorbed |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | PE |
| Host Species | Goat |
| Quantity | 1 mL |
| Primary or Secondary | Secondary |
| Target Species | Rat |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.