Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti- Rat IgG, H + L Chain Specific, Fluorescein, Secondary Antibody,

Catalog No. 4014142ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
Conjugate:
Fluorescein
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-141-42ML 2 mL Fluorescein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-141-42ML Supplier MilliporeSigma™ Supplier No. 4014142ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Specifically detects IgG, H & L Chain Specific Clone: in Rat samples, and it is validated for Immunoelectrophoresis, Immunofluorescence, Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG, H & L Chain Specific
Applications Immunoelectrophoresis, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Fluorescein
Formulation In 150mM NaCl, 20mM phosphate buffer, 1% BSA, pH 7.6.
Host Species Goat
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage −20°C
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.