Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG, IgA and IgM whole molecules, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89139077 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-077 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-077 Supplier Abnova Corporation Supplier No. PAB9420
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Antigen Goat anti-Rat IgG, IgA and IgM whole molecules
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat anti-rat IgG recognizes rat IgG IgA IgM whole molecules. This secondary antibody was purified using antigen affinity chromatography.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM Na2B4O7, 75mM NaCl, 5mM EDTA, pH 8.0 (0.01% sodium azide)
Host Species Goat
Immunogen Rat IgG, IgA and IgM whole molecules
Purification Method This product was prepared from polyspecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Quantity 2 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.