Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Rat IgG2b Recombinant Secondary Antibody, Alexa Fluor™ Plus 680

Catalog No. 5000102327
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-001-02327 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-001-02327 Supplier Invitrogen™ Supplier No. SA510377AFP680
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Goat Recombinant Monoclonal Secondary Antibody

Goat anti-Rat IgG2b, Recombinant Secondary Antibody specifically detects the Fc portion of rat IgG2b isotype and shows strong reactivity to total rat IgG. It has minimal to no cross-reactivity towards other rat IgG isotypes and immunoglobulins tested from other species. Alexa Fluor™ Plus conjugated recombinant antibodies provide bright signals and high signal-to-noise for your critical experiments. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG2b
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10H38L66
Concentration 1 mg/mL
Conjugate Alexa Fluor Plus 680
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.3-7.3
Host Species Goat
Immunogen Purified Rat IgG.
Purification Method Affinity chromatography
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.