Learn More
Bioworld Goat Anti-Rat IgG (H+L) Antibody (Polyclonal IgG), 1 mL
Supplier:  Bioworld 203200221
Encompass_Preferred
Goat Anti-Rat IgG (H+L) Antibody (Polyclonal IgG)Cat#20320022DetailsHost:GoatIsotype:IgGClass:PolyclonalImmunogen:Rat IgG (H+L)Conjugate:UnconjugatedForm InformationForm:LiquidConcentration:1 mg/mLPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:Shipped at 4°C. Upon delivery aliquot and store at -20°C. Avoid freeze / thaw cycles.Usage RecommendationsWestern Blot (WB):1:3000-1:5000ELISA:1:150-1:400Immunoprecipitation (IP):1:350 - 1:500
Catalog No. 50-198-341