Learn More
Description
This antibody may cross react with IgM from other species.
Specifications
Specifications
| Antigen | Rat IgM |
| Applications | ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Rat IgM. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.