Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light chain Goat anti-Rat, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP202BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP202BMI 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP202BMI Supplier MilliporeSigma Supplier No. AP202B
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Goat anti-Rat light chain Antibody, biotin conjugate is validated for use in ELISA, flow cytometry, western blot, immunocytochemistry, immunohistochemistry for the detection of light chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Rat light chain
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation Purified by immunoaffinity chromatography. Liquid in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 0.05% Sodium Azide. 15mg/mL BSA as stabilizer.
Host Species Goat
Immunogen purified rat IgG light chain.
Purification Method Affinity Purified
Quantity 0.5 mL
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Protect from light
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.