Learn More
Vector Laboratories Goat Anti-Human IgG Antibody Unconjugated 1.5 mg
SDP
Supplier:  Vector Laboratories AI30001.5
Encompass
These Goat Anti-Human IgG antibodies are purified by an affinity chromatography procedure to remove any low affinity antibodies, then cross-reactive antibodies are removed by solid-phase adsorption. The final product is subjected to rigorous quality control assays including immunodiffusion, solid-phase enzyme immunoassays, gel electrophoresis and solid-phase binding assays. These antibodies will recognize both heavy and light chains. Goat Anti-Human IgG Antibody is supplied in carrier-free, lyophilized form. After resuspension, it's ready for iodination, fluorochrome labeling, or enzyme conjugations. It can also be employed as a capture antibody in enzyme immunoassays or in other assays requiring carrier-free immunoglobulins.
Catalog No. NC0979636