Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Antiserum to Swine Serum (Forensic), MP Biomedicals™

Catalog No. MP0856007 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0856007 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0856007 Supplier MP Biomedicals, Inc Supplier No. 0856007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Antiserum to Swine Serum (Forensic)

  • Product is the lyophilized powder of goat antiserum to swine serum and buffer salts.
  • The antiserum is prepared by immunizing goats with swine serum; antiserum is delipidated and made specific by immunoadsorption with soluble antigens or solid-phase immunoadsorption where appropriate
  • This product is intended for use as a precipitating antibody. It gives sharp precipitin lines in immunoelectrophoresis, capillary tube, and immunodiffusion technique
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry
Classification Polyclonal
Concentration Total Protein: 59.9mg/mL
Conjugate Unconjugated
Description Product is the lyophilized powder ofgoat antiserum to swine serum and buffer salts.
Formulation Lyophilized PBS with 0.05% Sodium Azide
Host Species Goat
Purification Method solid-phase absorption
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute product with 2.0mL of deionized or distilled water.gentle swirling may be used to speedrehydration. Avoid vigorous shaking of the reconstituted material. If a small amount of precipitate is present, or if preparation appears cloudy, clarify by centrifugation or filtration.
Target Species Swine
Content And Storage Store product at 2-8°C. Store reconstituted material no longer than 4 weeks at 2-8°C. For long-term storage, aliquot and freeze reconstituted product at -20°C or lower
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.