Learn More
Biomatik Corporation Goat Bone Morphogenetic Protein 4 (BMP4) CLIA Kit
Supplier:  Biomatik Corporation EKU09938
Encompass_Preferred
Goat Bone Morphogenetic Protein 4 (BMP4) CLIA Kit for detecting BMP4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 12.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: A0A2Z4FRB3. Gene ID: 100860789. Target Synonyms: BMP2B; BMP2-B; BMP2B1; ZYME; DVR4; Bone morphogenetic protein 2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7113