Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat F(ab')2 Fragment To Human IgG (Whole Molecule) Secondary Polyclonal Antibody, MP Biomedicals

Catalog No. ICN55049 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55049 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55049 Supplier MP Biomedicals, Inc Supplier No. 0855049
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Host Species: Goat
Species Reactivity: Human

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG, Whole Molecule
Classification Polyclonal
Conjugate Unconjugated
Host Species Goat
Quantity 2 mL
Primary or Secondary Secondary
Target Species Human
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.