Learn More
American Research Products Inc Goat follicle stimulating horm
Catalog No. 5000329905
Encompass
Goat follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 20 mIU/mL-800 mIU/mL Sensitivity 20 mIU/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29905 Supplier American Research Products Inc Supplier No. CSBE13277G
Goat follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 20 mIU/mL-800 mIU/mL Sensitivity 20 mIU/mL Sample serum plasma Method Competitive