Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat Gamma Globulin

Catalog No. PI31871 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31871 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31871 Supplier Invitrogen™ Supplier No. 31871
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Storage: After opening, dilute only enough antibody for a single day's use. Country of Origin: USA.

Goat IgG, also known as goat immunoglobulin G, is an antibody that is produced by the immune system of goats in response to the presence of antigens. IgG is a type of immunoglobulin that plays a role in the recognition and neutralization of pathogens, such as bacteria and viruses. It binds to specific antigens and triggers various immune responses, including phagocytosis and complement activation. Goat IgG can be used in research to study the immune response to pathogens, as well as to develop diagnostic tests and therapeutic interventions for infectious diseases.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Liquid
Product Type Goat Gamma Globulin
Quantity 10 mg
Concentration 11.6 mg/mL
Formulation PBS with no preservative; pH 7.6
For Use With (Application) Control, ChIP Assay, Immunoprecipitation, Flow Cytometry
Isotype Ig
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.