Learn More
Biomatik Corporation Goat Growth Hormone Releasing Hormone (GHRH) ELISA Kit
Supplier:  Biomatik Corporation EKU04616
Encompass_Preferred
Goat Growth Hormone Releasing Hormone (GHRH) ELISA Kit for detecting GHRH in serum, plasma, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.49pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P63293. Target Synonyms: GRF; GHRF; Somatocrinin; Growth-Hormone-Releasing Factor; Sermorelin; Somatoliberin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8942