Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat IgG Fraction To Mouse IgG Fc Polyclonal Secondary Antibody, HRP, MP Biomedicals

Catalog No. ICN55554 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55554 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55554 Supplier MP Biomedicals, Inc Supplier No. 0855554
Add to Cart
Add to Cart

Host Species: Goat
Isotype: IgG
Species Reactivity: Mouse

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc, gamma-specific
Classification Polyclonal
Conjugate HRP
Host Species Goat
Quantity 2 mL
Primary or Secondary Secondary
Target Species Mouse
Form Purified
Isotype IgG
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.