Learn More
Innovative Research Inc Goat IgG Fractionated Purified 1gm
Supplier:  Innovative Research Inc IGTIGGGF1GM
Encompass_Preferred
Goat IgG Fractionated Purified from Innovative Research has been purified from goat serum by fractionation. This is a frozen liquid buffered in 10mM sodium phosphate, 150mM sodium chloride pH 7.2, with a concentration of 10mg/ml and a purity of greater than 97% as determined by SDS-PAGE analysis.
Catalog No. 50-203-6565