Learn More
STEMCELL Technologies Goat Anti-Mouse IgG (H+L) Antibody, Polyclonal, iFluor 647, Size: 200 ug
SDP
Supplier:  STEMCELL Technologies 1001081
Encompass
This goat polyclonal antibody reacts with the heavy chains on mouse IgG and the light chains common in most mouse immunoglobulins. Use the polyclonal goat anti-mouse IgG (HL) antibody with applications such as immunohistochemistry (IHC) and immunofluorescence (IF) assays. It has also been verified for use with immunocytochemistry (ICC) and flow cytometry (FC). This antibody has minimal cross-reactivity to human cow horse rabbit and pig serum proteins but may react to immunoglobulins from other species. Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0223