Learn More
American Research Products Inc Goat insulin like growth facto
Catalog No. 5000328547
Encompass
Goat insulin-like growth factor 1 (IGF-1) ELISA Kit from CUSABIO Detection Range 62 5 ng/ml-1000 ng/ml Sensitivity 12 5 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28547 Supplier American Research Products Inc Supplier No. CSBE15909G
Goat insulin-like growth factor 1 (IGF-1) ELISA Kit from CUSABIO Detection Range 62 5 ng/ml-1000 ng/ml Sensitivity 12 5 ng/ml Sample serum plasma tissue homogenates Method Sandwich