Learn More
Biomatik Corporation Goat Interleukin 1 Alpha (IL1a) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU08270
Encompass_Preferred
Goat Interleukin 1 Alpha (IL1a) High-sensitive ELISA Kit for detecting IL1a in serum, plasma, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 0.781-50pg/ml. Sensitivity: 0.32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P79161. Target Synonyms: IL1-A; IL-1α; IL1F1; Preinterleukin 1 Alpha; Hematopoietin-1; Pro-Interleukin-1-Alpha; Interleukin-1 Family Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8683