Learn More
American Research Products Inc Goat interleukin 12 IL 12 P40
Catalog No. 5000329795
Encompass
Goat interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 9 37 pg/mL-600 pg/mL Sensitivity 2 34 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29795 Supplier American Research Products Inc Supplier No. CSBE12103G
Goat interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 9 37 pg/mL-600 pg/mL Sensitivity 2 34 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich