Learn More
Cusabio Technology LLC Goat Interleukin 4IL-4 ELISA
Catalog No. 5000050624
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Goat Interleukin 4 (IL-4) in serum plasma and tissue homogenates using a sandwich assay method The kit has a detection range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL ensuring precise measurement of low-abundance targets Results can be obtained within 1-5 hours using a sample volume of 50-100 L per test The assay utilizes a 450 nm wavelength for optical measurement providing reliable and reproducible data for research applications Suitable for laboratories requiring accurate IL-4 quantification this kit combines efficiency with robust performance across biological sample types
Catalog No. 50-000-50624 Supplier Cusabio Technology LLC Supplier No. CSBE09808G
This ELISA kit is designed for the quantitative detection of Goat Interleukin 4 (IL-4) in serum plasma and tissue homogenates using a sandwich assay method The kit has a detection range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL ensuring precise measurement of low-abundance targets Results can be obtained within 1-5 hours using a sample volume of 50-100 L per test The assay utilizes a 450 nm wavelength for optical measurement providing reliable and reproducible data for research applications Suitable for laboratories requiring accurate IL-4 quantification this kit combines efficiency with robust performance across biological sample types