Learn More
Cusabio Technology LLC Goat Interleukin 6IL-6 ELISA
Catalog No. 5000051190
Encompass_Preferred
This ELISA kit is designed for the detection of Goat Interleukin 6 (IL-6) in various biological samples including serum plasma and tissue homogenates Utilizing a quantitative sandwich assay principle it offers a sensitive detection range from 1 56 pg/mL to 100 pg/mL with a minimum detectable concentration of 0 39 pg/mL The kit requires a short incubation period of 1-5 hours and operates with a sample volume of 50-100 L per test Measurements are performed at an absorbance wavelength of 450 nm ensuring accurate and reliable quantification of IL-6 levels Its standardized protocol and consistent performance make it suitable for research applications requiring precise monitoring of interleukin-6 concentrations in goat-derived specimens
Catalog No. 50-000-51190 Supplier Cusabio Technology LLC Supplier No. CSBE14360G
This ELISA kit is designed for the detection of Goat Interleukin 6 (IL-6) in various biological samples including serum plasma and tissue homogenates Utilizing a quantitative sandwich assay principle it offers a sensitive detection range from 1 56 pg/mL to 100 pg/mL with a minimum detectable concentration of 0 39 pg/mL The kit requires a short incubation period of 1-5 hours and operates with a sample volume of 50-100 L per test Measurements are performed at an absorbance wavelength of 450 nm ensuring accurate and reliable quantification of IL-6 levels Its standardized protocol and consistent performance make it suitable for research applications requiring precise monitoring of interleukin-6 concentrations in goat-derived specimens