Learn More
R&D Systems™ Goat Keratan Sulfate Proteoglycans Protein
Description
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Molecular Weight (g/mol) | Observed M.W.: 30-300kDa, reducing conditions, Theoretical M.W.: |
| Quantity | 10 μg |
| Source | Goat cornea-derived Keratan Sulfate Proteoglycans protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU per 1μg of the protein by the LAL method. |
| Common Name | Keratan Sulfate Proteoglycans |
| Biological Activity | Measured by its ability to act as a substrate for Recombinant F. keratolyticus Endo-beta-galactosidase. >90% of Keratan Sulfate Proteoglycans can be cleaved under the described conditions. See Activity Assay Protocol on RnDSystems.com. |
| Conjugate | Unconjugated |
| Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.