Learn More
American Research Products Inc Goat Luteinizing Hormone LH
Catalog No. 5000329838
Encompass
Goat Luteinizing Hormone (LH) ELISA KIT from CUSABIO Detection Range 0 3 mLU/mL-60 mLU/mL Sensitivity 0 15 mIU/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29838 Supplier American Research Products Inc Supplier No. CSBE13274G
Goat Luteinizing Hormone (LH) ELISA KIT from CUSABIO Detection Range 0 3 mLU/mL-60 mLU/mL Sensitivity 0 15 mIU/mL Sample serum plasma tissue homogenates Method Competitive