Learn More
Biomatik Corporation Goat Luteinizing Hormone (LH) ELISA Kit
Supplier:  Biomatik Corporation EKU05692
Encompass_Preferred
Goat Luteinizing Hormone (LH) ELISA Kit for detecting LH in serum, plasma, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 370.4-30000pg/ml. Sensitivity: 157.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8WMW8; Q1RP71. Gene ID: 100860817; 102180531. Target Synonyms: ICSH; Lutropin; Interstitial Cell Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8890