Learn More
Cayman Chemical Goat ntI-GST IgG SureLIght1ea
SDP
Catalog No. 502953819
Encompass_Preferred
Goat anti-GST (Glutathione-S-Transferase) IgG conjugated to SureLight™ P3 Antibody: Goat polyclonal anti-Glutathione-S-Transferase (GST) Dye: SureLight™ P3 Excitation max. λ: 614 nm Emission max. λ: 662 nm Isotype: IgG P3
Catalog No. 50-295-3819 Supplier Cayman Chemical Supplier No. 134481 EA
Goat anti-GST (Glutathione-S-Transferase) IgG conjugated to SureLight™ P3 Antibody: Goat polyclonal anti-Glutathione-S-Transferase (GST) Dye: SureLight™ P3 Excitation max. λ: 614 nm Emission max. λ: 662 nm Isotype: IgG P3