Learn More
Cayman Chemical Goat ntIGST IgG DyLIght 55 1ea
SDP
Catalog No. 502951140
Encompass_Preferred
Goat anti-GST (Glutathione-S-Transferase) IgG conjugated to DyLight 550 Antibody: Goat polyclonal anti-GST IgG Dye: DyLight 550 Excitation max. λ: 562 nm Emission max. λ: 576 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays
Catalog No. 50-295-1140 Supplier Cayman Chemical Supplier No. 112541 EA
Goat anti-GST (Glutathione-S-Transferase) IgG conjugated to DyLight 550 Antibody: Goat polyclonal anti-GST IgG Dye: DyLight 550 Excitation max. λ: 562 nm Emission max. λ: 576 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays