Learn More
Cayman Chemical Goat ntIGST IgG SureLIght 1ea
SDP
Catalog No. 502951115
Encompass_Preferred
Goat anti-GST (Glutathione-S-Transferase) IgG conjugated to SureLight PerCP Antibody: Goat polyclonal anti-GST IgG Dye: SureLight™ Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, Protein microarrays
Catalog No. 50-295-1115 Supplier Cayman Chemical Supplier No. 112721 EA
Goat anti-GST (Glutathione-S-Transferase) IgG conjugated to SureLight PerCP Antibody: Goat polyclonal anti-GST IgG Dye: SureLight™ Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, Protein microarrays