Learn More
Cayman Chemical Goat ntIHumn IgA SureLIght
SDP
Catalog No. 502965301
Encompass_Preferred
Goat Anti-Human IgA conjugated to SureLight™ R-PE, 100μg Antibody: Goat polyclonal Anti-Human IgA Isotype: IgA Uses: Flow cytometry, Fluorescence-activated cell sorting, Luminex, and Protein Microarrays
Catalog No. 50-296-5301 Supplier Cayman Chemical Supplier No. 16639100 UG
Goat Anti-Human IgA conjugated to SureLight™ R-PE, 100μg Antibody: Goat polyclonal Anti-Human IgA Isotype: IgA Uses: Flow cytometry, Fluorescence-activated cell sorting, Luminex, and Protein Microarrays