Learn More
Cayman Chemical Goat ntIHumn IgG FcSpec H 1mg
SDP
Catalog No. 502965331
Encompass_Preferred
Goat anti-Human IgG, Fc-specific conjugated to HRP Antibody: Goat polyclonal anti-Human IgG Isotype: IgG Enzyme: Horseradish Peroxidase Uses: ELISA and Western blot detection. Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000.
Catalog No. 50-296-5331 Supplier Cayman Chemical Supplier No. 167371 MG
Goat anti-Human IgG, Fc-specific conjugated to HRP Antibody: Goat polyclonal anti-Human IgG Isotype: IgG Enzyme: Horseradish Peroxidase Uses: ELISA and Western blot detection. Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000.