Learn More
Cayman Chemical Goat ntIHumn IgM SureLIght
SDP
Catalog No. 502966177
Encompass_Preferred
Goat Anti-Human IgM conjugated to SureLight™ APC, 100UG Antibody: Goat polyclonal Anti-human IgM Uses: Optimized for High throughput screening (HTS) and Fluorescence resonance energy transfer (FRET) assays, Flow cytometry, Microscopy
Catalog No. 50-296-6177 Supplier Cayman Chemical Supplier No. 16586100 UG
Goat Anti-Human IgM conjugated to SureLight™ APC, 100UG Antibody: Goat polyclonal Anti-human IgM Uses: Optimized for High throughput screening (HTS) and Fluorescence resonance energy transfer (FRET) assays, Flow cytometry, Microscopy