Learn More
Cayman Chemical Goat ntIlama IgG SureLIght 1ea
SDP
Catalog No. 502953816
Encompass_Preferred
Goat anti-Llama IgG conjugated to SureLight APC, 100μg Antibody: Goat polyclonal anti-Llama IgG Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry/Fluorescence-activated cell sorting
Catalog No. 50-295-3816 Supplier Cayman Chemical Supplier No. 133931 EA
Goat anti-Llama IgG conjugated to SureLight APC, 100μg Antibody: Goat polyclonal anti-Llama IgG Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry/Fluorescence-activated cell sorting