Learn More
Cayman Chemical Goat ntIMouse IgE SureLIght
SDP
Catalog No. 502965118
Encompass_Preferred
Goat anti-Mouse IgE conjugated to SureLight™ R-PE Antibody: Goat polyclonal anti-Mouse IgE Isotype: IgE Dye: SureLight™ R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Uses: Flow cytometry, Fluorescence-activated cell sorting, Luminex, and Protein Microarrays
Catalog No. 50-296-5118 Supplier Cayman Chemical Supplier No. 16646100 UG
Goat anti-Mouse IgE conjugated to SureLight™ R-PE Antibody: Goat polyclonal anti-Mouse IgE Isotype: IgE Dye: SureLight™ R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Uses: Flow cytometry, Fluorescence-activated cell sorting, Luminex, and Protein Microarrays