Learn More
Biomatik Corporation Goat Prolactin (PRL) ELISA Kit
Supplier:  Biomatik Corporation EKU11365
Encompass_Preferred
Goat Prolactin (PRL) ELISA Kit for detecting PRL in serum, plasma, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28318. Gene ID: 100861193. Target Synonyms: LTH; Luteotropic Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8157